Ergosterol, 96%, may cont. up to ca 6% water
Ergosterol, 96%, may cont. up to ca 6% water
Ergosterol, 96%, may cont. up to ca 6% water
Thermo Scientific Chemicals

Ergosterol, 96%, may cont. up to ca 6% water

Store cold | CAS: 57-87-4 | C28H44O | 396.659 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23840.06
also known as B23840-06
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.80
Save 12.15 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.80
Save 12.15 (15%)
Each
Chemical Identifiers
CAS57-87-4
IUPAC Name(1R,3aR,7S,9aR,9bS,11aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC28H44O
InChI KeyDNVPQKQSNYMLRS-APGDWVJJSA-N
SMILESCC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-120? ? 15? (c=1 in Chloroform)
FormCrystals or powder or crystalline powder or needles
Appearance (Color)White to cream
Assay (HPLC)≥95.0%
Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

Solubility
Soluble in water (<1mg/mL at 25°C), chloroform (50 mg/ml), DMSO (<1mg/mL at 25°C), ethanol (<1mg/mL at 25°C), and ether (14.29 mg/ml).

Notes
Light sensitive. Store away from air, light and oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark. Store under inert gas.
RUO – Research Use Only

General References:

  1. H. Van Den Bossche, et al. Biochemical effects of miconazole on fungi. II. Inhibition of ergosterol biosynthesis in Candida albicans.Chem. Biol. Interact.,1978,21(1), 59-78.
  2. F. Martin, et al. An improved ergosterol assay to estimate fungal biomass in ectomycorrhizas.Mycological Research.,1990,94(8), 1059-1064.