Furan-2-boronic acid, 97%
Furan-2-boronic acid, 97%
Furan-2-boronic acid, 97%
Furan-2-boronic acid, 97%
Thermo Scientific Chemicals

Furan-2-boronic acid, 97%

CAS: 13331-23-2 | C4H5BO3 | 111.891 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23842.06
also known as B23842-06
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS13331-23-2
IUPAC Name(furan-2-yl)boronic acid
Molecular FormulaC4H5BO3
InChI KeyPZJSZBJLOWMDRG-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
Appearance (Color)Pale cream to cream
It is used in Suzuki reaction for generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in Suzuki reaction for generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Michael J. Burns, et al. Simple Palladium(II) Precatalyst for Suzuki-Miyaura Couplings: Efficient Reactions of Benzylic, Aryl, Heteroaryl, and Vinyl Coupling Partners.Org. Lett.,2007,9(26), 5397-5400.
  2. Khokan Samanta, et al. A generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes through Suzuki reactions.Tetrahedron Lett.,2008,49(9), 1461-1464.
  3. Undergoes an asymmetric boronic acid Mannich reaction with aldehydes in the presence of the chiral template (S)-5-phenylmorpholine-2-one to give substituted furfurylamines with high diastereomeric excess: Chem. Commun., 1953 (1996); J. Chem. Soc., Perkin 1, 2982 (2000):