3,5-Dibromobenzeneboronic acid, 97%
3,5-Dibromobenzeneboronic acid, 97%
3,5-Dibromobenzeneboronic acid, 97%
Thermo Scientific Chemicals

3,5-Dibromobenzeneboronic acid, 97%

CAS: 117695-55-3 | C6H5BBr2O2 | 279.722 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23863.06
also known as B23863-06
Price (USD)
90.65
Online Exclusive
101.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
90.65
Online Exclusive
101.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS117695-55-3
IUPAC Name(3,5-dibromophenyl)boronic acid
Molecular FormulaC6H5BBr2O2
InChI KeyWQBLCGDZYFKINX-UHFFFAOYSA-N
SMILESOB(O)C1=CC(Br)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0%
Appearance (Color)Pale cream to cream to yellow
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only