Isophthalonitrile, 97+%
Isophthalonitrile, 97+%
Isophthalonitrile, 97+%
Thermo Scientific Chemicals

Isophthalonitrile, 97+%

CAS: 626-17-5 | C8H4N2 | 128.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B23900.30
also known as B23900-30
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.00
Save 7.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.00
Save 7.35 (15%)
Each
Chemical Identifiers
CAS626-17-5
IUPAC Namebenzene-1,3-dicarbonitrile
Molecular FormulaC8H4N2
InChI KeyLAQPNDIUHRHNCV-UHFFFAOYSA-N
SMILESN#CC1=CC(=CC=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Assay (GC)≥97.0%
FormPowder or flakes
Melting Point (clear melt)158-165?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only