2,4-Bis(trifluoromethyl)benzyl bromide, 97%
2,4-Bis(trifluoromethyl)benzyl bromide, 97%
2,4-Bis(trifluoromethyl)benzyl bromide, 97%
2,4-Bis(trifluoromethyl)benzyl bromide, 97%
Thermo Scientific Chemicals

2,4-Bis(trifluoromethyl)benzyl bromide, 97%

CAS: 140690-56-8 | C9H5BrF6 | 307.033 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23910.06
also known as B23910-06
Price (USD)
234.65
Special offer
Online exclusive
Ends: 30-Jun-2026
276.00
Save 41.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
234.65
Special offer
Online exclusive
Ends: 30-Jun-2026
276.00
Save 41.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS140690-56-8
IUPAC Name1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
Molecular FormulaC9H5BrF6
InChI KeySWFFFUJOWAJJCH-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(CBr)C(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4480-1.4530 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only