2,3-Dimethylbenzeneboronic acid, 98%
2,3-Dimethylbenzeneboronic acid, 98%
2,3-Dimethylbenzeneboronic acid, 98%
Thermo Scientific Chemicals

2,3-Dimethylbenzeneboronic acid, 98%

CAS: 183158-34-1 | C8H11BO2 | 149.984 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23942.06
also known as B23942-06
Price (USD)
76.65
Online Exclusive
84.80
Save 8.15 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
76.65
Online Exclusive
84.80
Save 8.15 (10%)
Each
Chemical Identifiers
CAS183158-34-1
IUPAC Name(2,3-dimethylphenyl)boronic acid
Molecular FormulaC8H11BO2
InChI KeyZYYANAWVBDFAHY-UHFFFAOYSA-N
SMILESCC1=CC=CC(B(O)O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)174.0-180.0?C
Proton NMRConforms to structure
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only