2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid, 97%
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid, 97%
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid, 97%
Thermo Scientific Chemicals

2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid, 97%

CAS: 126120-85-2 | C8H4F2O4 | 202.113 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23955.03
also known as B23955-03
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.00
Save 7.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.00
Save 7.35 (15%)
Each
Chemical Identifiers
CAS126120-85-2
IUPAC Name2,2-difluoro-2H-1,3-benzodioxole-4-carboxylic acid
Molecular FormulaC8H4F2O4
InChI KeyZGAQVJDFFVTWJK-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC2=C1OC(F)(F)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)198.0-208.0?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only