2-(Bromoacetyl)naphthalene, 98%
2-(Bromoacetyl)naphthalene, 98%
2-(Bromoacetyl)naphthalene, 98%
Thermo Scientific Chemicals

2-(Bromoacetyl)naphthalene, 98%

CAS: 613-54-7 | C12H9BrO | 249.107 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B23960.09
also known as B23960-09
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.70
Save 11.05 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.70
Save 11.05 (15%)
Each
Chemical Identifiers
CAS613-54-7
IUPAC Name2-bromo-1-(naphthalen-2-yl)ethan-1-one
Molecular FormulaC12H9BrO
InChI KeyYHXHHGDUANVQHE-UHFFFAOYSA-N
SMILESBrCC(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink to pink to purple to brown
Assay (GC)≥97.5%
Melting Point (clear melt)79.5-85.5?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only