Benzenesulfonyl hydrazide, 98%
Benzenesulfonyl hydrazide, 98%
Benzenesulfonyl hydrazide, 98%
Thermo Scientific Chemicals

Benzenesulfonyl hydrazide, 98%

CAS: 80-17-1 | C6H8N2O2S | 172.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23971.14
also known as B23971-14
Price (USD)
52.65
Online Exclusive
58.10
Save 5.45 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
52.65
Online Exclusive
58.10
Save 5.45 (9%)
Each
Chemical Identifiers
CAS80-17-1
IUPAC Namebenzenesulfonohydrazide
Molecular FormulaC6H8N2O2S
InChI KeyVJRITMATACIYAF-UHFFFAOYSA-N
SMILESNNS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥97.5% (based on carbon EA)
FormPowder or crystals
Elemental AnalysisNitrogen N: 15.86-16.68% (theory 16.27%)
Appearance (Color)White to pale cream
Elemental AnalysisCarbon C: 40.80-42.90% (theory 41.85%)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only