2,2'-Furil dioxime, mixture of isomers, 97%
2,2'-Furil dioxime, mixture of isomers, 97%
2,2'-Furil dioxime, mixture of isomers, 97%
Thermo Scientific Chemicals

2,2'-Furil dioxime, mixture of isomers, 97%

CAS: 522-27-0 | C10H8N2O4 | 220.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23972.06
also known as B23972-06
Price (USD)
287.65
Special offer
Online exclusive
Ends: 30-Jun-2026
338.00
Save 50.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
287.65
Special offer
Online exclusive
Ends: 30-Jun-2026
338.00
Save 50.35 (15%)
Each
Chemical Identifiers
CAS522-27-0
IUPAC Name(E)-N-[(2Z)-1,2-bis(furan-2-yl)-2-(hydroxyimino)ethylidene]hydroxylamine
Molecular FormulaC10H8N2O4
InChI KeyRBOZTFPIXJBLPK-DSOJMZEYSA-N
SMILESO\N=C(\C(=N/O)\C1=CC=CO1)/C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)Pale cream to cream to yellow to pale brown
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only