Triacetoxy(methyl)silane, tech. 90%
Triacetoxy(methyl)silane, tech. 90%
Triacetoxy(methyl)silane, tech. 90%
Thermo Scientific Chemicals

Triacetoxy(methyl)silane, tech. 90%

CAS: 4253-34-3 | C7H12O6Si | 220.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B23974.30
also known as B23974-30
Price (USD)
368.65
Special offer
Online exclusive
Ends: 30-Jun-2026
434.00
Save 65.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
368.65
Special offer
Online exclusive
Ends: 30-Jun-2026
434.00
Save 65.35 (15%)
Each
Chemical Identifiers
CAS4253-34-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or clear colorless to pale yellow
FormFused solid or liquid as melt
Assay (Aqueous acid-base Titration)≥88.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only