1-Bromo-4-n-butylbenzene, 98%
1-Bromo-4-n-butylbenzene, 98%
1-Bromo-4-n-butylbenzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-n-butylbenzene, 98%

CAS: 41492-05-1 | C10H13Br | 213.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B23979.14
also known as B23979-14
Price (USD)
72.65
Online Exclusive
80.00
Save 7.35 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.00
Save 7.35 (9%)
Each
Chemical Identifiers
CAS41492-05-1
IUPAC Name1-bromo-4-butylbenzene
Molecular FormulaC10H13Br
InChI KeyBRGVKVZXDWGJBX-UHFFFAOYSA-N
SMILESCCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5285-1.5325 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only