N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
Thermo Scientific Chemicals

N,N-Dimethylethylamine, 99%

CAS: 598-56-1 | C4H11N | 73.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number B23992.AE
also known as B23992-AE
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.80
Save 7.15 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.80
Save 7.15 (15%)
Each
Chemical Identifiers
CAS598-56-1
IUPAC Nameethyldimethylamine
Molecular FormulaC4H11N
InChI KeyDAZXVJBJRMWXJP-UHFFFAOYSA-N
SMILESCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.3715-1.3745 @ 20?C
N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents and peroxide.
RUO – Research Use Only

General References:

  1. Chen, J.; Wang, Y.; Huang, Y.; Xu, K.; Li, N.; Wen, Q.; Zhou, Y. Magnetic multiwall carbon nanotubes modified with dual hydroxy functional ionic liquid for the solid-phase extraction of protein. Analyst 2015, 140 (10), 3474-3483.
  2. Wunderlich, C. H.; Juen, M. A.; LeBlanc, R. M.; Longhini, A. P.; Dayie, T. K.; Kreutz, C. Chapter Eighteen - Stable Isotope-Labeled RNA Phosphoramidites to Facilitate Dynamics by NMR. Methods Enzymol. 2015, 565, 461-494.