3,6-Di-tert-butylfluorene, 98%
3,6-Di-tert-butylfluorene, 98%
3,6-Di-tert-butylfluorene, 98%
Thermo Scientific Chemicals

3,6-Di-tert-butylfluorene, 98%

CAS: 58775-07-8 | C21H26 | 278.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23996.06
also known as B23996-06
Price (USD)
394.65
Online Exclusive
438.00
Save 43.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
394.65
Online Exclusive
438.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS58775-07-8
IUPAC Name3,6-di-tert-butyl-9H-fluorene
Molecular FormulaC21H26
InChI KeyIWUHFBNEZZKUEY-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC2=C(CC3=C2C=C(C=C3)C(C)(C)C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)77.0-83.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only