Thermo Scientific Chemicals

Dichloromethylphenylsilane, 98%, Thermo Scientific Chemicals

Catalog number: B23999.14
25 g, Each
Thermo Scientific Chemicals

Dichloromethylphenylsilane, 98%, Thermo Scientific Chemicals

Catalog number: B23999.14
25 g, Each
Quantity
Catalog number: B23999.14
also known as B23999-14
Price (USD)
Price: 25.30
Online price: 21.65
Your price:
Quantity
-

Chemical Identifiers

CAS
149-74-6
IUPAC Name
dichloro(methyl)phenylsilane
Molecular Formula
C7H8Cl2Si
InChI Key
GNEPOXWQWFSSOU-UHFFFAOYSA-N
SMILES
C[Si](Cl)(Cl)C1=CC=CC=C1
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5170-1.5210 @ 20?C
Appearance (Color)
Clear colorless to pale pink
Identification (FTIR)
Conforms

Description

Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is sensitive to moisture. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text