Isovaleryl chloride, 97%
Isovaleryl chloride, 97%
Isovaleryl chloride, 97%
Thermo Scientific Chemicals

Isovaleryl chloride, 97%

CAS: 108-12-3 | C5H9ClO | 120.576 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B24002.14
also known as B24002-14
Price (USD)
30.65
Online Exclusive
33.40
Save 2.75 (8%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
33.40
Save 2.75 (8%)
Each
Chemical Identifiers
CAS108-12-3
IUPAC Name3-methylbutanoyl chloride
Molecular FormulaC5H9ClO
InChI KeyISULZYQDGYXDFW-UHFFFAOYSA-N
SMILESCC(C)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.4135-1.4175 @ 20°C
Assay (GC)≥95.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only