Ethylurea, 98%
Ethylurea, 98%
Ethylurea, 98%
Thermo Scientific Chemicals

Ethylurea, 98%

CAS: 625-52-5 | C3H8N2O | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B24040.36
also known as B24040-36
Price (USD)
504.65
Online Exclusive
561.00
Save 56.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
504.65
Online Exclusive
561.00
Save 56.35 (10%)
Each
Chemical Identifiers
CAS625-52-5
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5%
Appearance (Color)White to pale yellow or cream
FormCrystalline powder
Mass SpecConsistent with structure
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.

Solubility
Soluble in methanol (25 mg/ml), and water (100 mg/ml).

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hideshi Suzuki; Hisakazu Sunada. Comparison of Nicotinamide, Ethylurea and Polyethylene Glycol as Carriers for Nifedipine Solid Dispersion Systems. Chemical and Pharmaceutical Bulletin. 1997, 45 (10), 1688-1693.
  2. Michael J. Capaldi and John A. Chapman. The C-terminal extrahelical peptide of type I collagen and its role in fibrillogenesis in vitro: Effects of Ethylurea. Biopolymers. 1984, 23 (2), 313-323.