2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%
Thermo Scientific Chemicals

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%

CAS: 722-92-9 | C9H7F6NO | 259.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24048.14
also known as B24048-14
Price (USD)
405.65
Online Exclusive
450.00
Save 44.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
405.65
Online Exclusive
450.00
Save 44.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS722-92-9
IUPAC Name2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC9H7F6NO
InChI KeyTZEJXCIGVMTMDY-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (GC)≥95.0%
FormPowder
Melting Point (clear melt)145.0-154.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only