1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%
1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%
1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%
1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%
Thermo Scientific Chemicals

1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%

CAS: 2043-55-2 | C6H4F9I | 373.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24075.14
also known as B24075-14
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2043-55-2
IUPAC Name1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
Molecular FormulaC6H4F9I
InChI KeyCXHFIVFPHDGZIS-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.3675-1.3725 @ 20°C
Appearance (Color)Clear colorless to pale yellow or pink
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only