Thermo Scientific Chemicals

4-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24117.14
25 g, Each
Thermo Scientific Chemicals

4-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24117.14
25 g, Each
Quantity
Catalog number: B24117.14
also known as B24117-14
Price (USD)
Price: 861.00
Online price: 744.65
Your price:
Quantity
-

Chemical Identifiers

CAS
139911-29-8
IUPAC Name
(4-fluoro-2-methylphenyl)boronic acid
Molecular Formula
C7H8BFO2
InChI Key
IQMLIVUHMSIOQP-UHFFFAOYSA-N
SMILES
CC1=CC(F)=CC=C1B(O)O
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5%
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text