1-Methylthio-2-propanone, 98+%
1-Methylthio-2-propanone, 98+%
1-Methylthio-2-propanone, 98+%
Thermo Scientific Chemicals

1-Methylthio-2-propanone, 98+%

CAS: 14109-72-9 | C4H8OS | 104.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24149.06
also known as B24149-06
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.50
Save 10.85 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.50
Save 10.85 (15%)
Each
Chemical Identifiers
CAS14109-72-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.4725-1.4765 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Paul G. Gassman.; Gordon Gruetzmacher.; T. J. Van Bergen. Generation of azasulfonium salts from halogen-sulfide complexes and anilines. Synthesis of indoles, oxindoles, and alkylated aromatic amines bearing cation stabilizing substituents. J. Am. Chem. Soc. 1974, 96 (17), 5512-5517.
  2. Paul G. Gassman.; David R. Amick. A Simple Route to Benzofurans. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 1975, 5 (5), 325-329.