2-Naphthaleneboronic acid, 97%
2-Naphthaleneboronic acid, 97%
2-Naphthaleneboronic acid, 97%
Thermo Scientific Chemicals

2-Naphthaleneboronic acid, 97%

CAS: 32316-92-0 | C10H9BO2 | 171.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24157.03
also known as B24157-03
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
30.60
Save 3.95
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
30.60
Save 3.95
Each
Chemical Identifiers
CAS32316-92-0
IUPAC Name(naphthalen-2-yl)boronic acid
Molecular FormulaC10H9BO2
InChI KeyKPTRDYONBVUWPD-UHFFFAOYSA-N
SMILESOB(O)C1=CC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Identification (FTIR)Conforms
Proton NMRConforms to structure
FormPowder or crystalline powder and/or chunks
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only