1,5,5-Trimethylhydantoin, 98%
1,5,5-Trimethylhydantoin, 98%
1,5,5-Trimethylhydantoin, 98%
Thermo Scientific Chemicals

1,5,5-Trimethylhydantoin, 98%

CAS: 6851-81-6 | C6H10N2O2 | 142.158 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24188.06
also known as B24188-06
Price (USD)
48.65
Online Exclusive
54.30
Save 5.65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
54.30
Save 5.65 (10%)
Each
Chemical Identifiers
CAS6851-81-6
IUPAC Name1,5,5-trimethylimidazolidine-2,4-dione
Molecular FormulaC6H10N2O2
InChI KeyZNYIPTYJBRGSSL-UHFFFAOYSA-N
SMILESCN1C(=O)NC(=O)C1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Melting Point (clear melt)159.5-165.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only