1-Bromo-2,3,5,6-tetrafluorobenzene, 98%
1-Bromo-2,3,5,6-tetrafluorobenzene, 98%
1-Bromo-2,3,5,6-tetrafluorobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-2,3,5,6-tetrafluorobenzene, 98%

CAS: 1559-88-2 | C6HBrF4 | 228.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24265.06
also known as B24265-06
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS1559-88-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4675-1.4715 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. GB Deacon and DG Vince. Symmetrization of Bromobis(polyfluorophenyl)thallium(III) compounds with copper powder. Australian Journal of Chemistry. 1975, 28 (9), 1931 - 1941.
  2. Zongjun Qiao; Hui Liu; Xiao Xiao; Yana Fu; Jianpeng Wei; Yuxue Li; and Xuefeng Jiang. Efficient Access to 1,4-Benzothiazine: Palladium-Catalyzed Double C-S Bond Formation Using Na2S2O3 as Sulfurating Reagent. Org. Lett. 2013, 15 (11), 2594-2597.