2,6-Dichloro-4-nitroaniline, 95%
2,6-Dichloro-4-nitroaniline, 95%
2,6-Dichloro-4-nitroaniline, 95%
Thermo Scientific Chemicals

2,6-Dichloro-4-nitroaniline, 95%

CAS: 99-30-9 | C6H4Cl2N2O2 | 207.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B24283.22
also known as B24283-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS99-30-9
IUPAC Name2,6-dichloro-4-nitroaniline
Molecular FormulaC6H4Cl2N2O2
InChI KeyBIXZHMJUSMUDOQ-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=C(C=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
Melting Point (clear melt)183.5-194.5?C
It is used as dyestuff, agricultural and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as dyestuff, agricultural and pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only