Ethyl 3-(dimethylamino)acrylate, 99%
Ethyl 3-(dimethylamino)acrylate, 99%
Ethyl 3-(dimethylamino)acrylate, 99%
Thermo Scientific Chemicals

Ethyl 3-(dimethylamino)acrylate, 99%

CAS: 924-99-2 | C7H13NO2 | 143.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B24291.09
also known as B24291-09
Price (USD)
46.65
Online exclusive
51.60
Save 4.95
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
46.65
Online exclusive
51.60
Save 4.95
Each
Chemical Identifiers
CAS924-99-2
IUPAC Nameethyl (2Z)-3-(dimethylamino)prop-2-enoate
Molecular FormulaC7H13NO2
InChI KeyMVUMJYQUKKUOHO-WAYWQWQTSA-N
SMILESCCOC(=O)C=C/N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5105-1.5135 @ 20?C
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water 90 g/L (25°C).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Violetta Cecchetti; Oriana Tabarrini; Stefano Sabatini; Hua Miao; Enrica Filipponi; Arnaldo Fravolini. Studies on 6-Aminoquinolones: synthesis and antibacterial evaluation of 6-amino-8-ethyl- and 6-amino-8-methoxyquinolones. Bioorganic & Medicinal Chemistry. 1999, 7 (11), 2465-2471.
  2. Feng V. Yang; William D. Shipe; Jaime L. Bunda; M. Brad Nolt; David D. Wisnoski; Zhijian Zhao; James C. Barrow; William J. Ray; Lei Ma; Marion Wittmann; Matthew A. Seager; Kenneth A. Koeplinger; George D. Hartman; Craig W. Lindsley. Parallel synthesis of N-biaryl quinolone carboxylic acids as selective M1 positive allosteric modulators. Bioorganic & Medicinal Chemistry Letters. 2010, 20 (2), 531-536.