3,4-Dichlorobenzeneboronic acid, 97%
3,4-Dichlorobenzeneboronic acid, 97%
3,4-Dichlorobenzeneboronic acid, 97%
Thermo Scientific Chemicals

3,4-Dichlorobenzeneboronic acid, 97%

CAS: 151169-75-4 | C6H5BCl2O2 | 190.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24292.03
also known as B24292-03
Price (USD)
16.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
16.00
Each
Chemical Identifiers
CAS151169-75-4
IUPAC Name(3,4-dichlorophenyl)boronic acid
Molecular FormulaC6H5BCl2O2
InChI KeyJKIGHOARKAIPJI-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only