Cobalt(II) gluconate hydrate
Thermo Scientific Chemicals

Cobalt(II) gluconate hydrate

CAS: 1809162-99-9 | C12H26CoO15 | 469.258 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B24293.14
also known as B24293-14
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
Chemical Identifiers
CAS1809162-99-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
FormPowder
Complexometric Titration97-103%
Trace MetalLead (Pb) : <10ppm
Trace MetalNickel (Ni) : <10ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only