2,6-Dimethoxybenzeneboronic acid, 97+%
2,6-Dimethoxybenzeneboronic acid, 97+%
2,6-Dimethoxybenzeneboronic acid, 97+%
Thermo Scientific Chemicals

2,6-Dimethoxybenzeneboronic acid, 97+%

CAS: 23112-96-1 | C8H11BO4 | 181.982 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24305.06
also known as B24305-06
Price (USD)
50.65
Online Exclusive
56.30
Save 5.65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.30
Save 5.65 (10%)
Each
Chemical Identifiers
CAS23112-96-1
IUPAC Name(2,6-dimethoxyphenyl)boronic acid
Molecular FormulaC8H11BO4
InChI KeyBKWVXPCYDRURMK-UHFFFAOYSA-N
SMILESCOC1=CC=CC(OC)=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.0%
Assay (HPLC)≥97.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only