N-Acetyl-L-methionine, 99%
N-Acetyl-L-methionine, 99%
N-Acetyl-L-methionine, 99%
N-Acetyl-L-methionine, 99%
Thermo Scientific Chemicals

N-Acetyl-L-methionine, 99%

CAS: 65-82-7 | C7H13NO3S | 191.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24312.06
also known as B24312-06
Price (USD)
46.65
Online Exclusive
51.60
Save 4.95 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.60
Save 4.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS65-82-7
IUPAC Name(2S)-2-acetamido-4-(methylsulfanyl)butanoic acid
Molecular FormulaC7H13NO3S
InChI KeyXUYPXLNMDZIRQH-LURJTMIESA-N
SMILESCSCC[C@H](NC(C)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Optical Rotation-20.5° to -22.5° (c=1.3 in water)
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)101.0-107.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only