2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%
Thermo Scientific Chemicals

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%

CAS: 651-83-2 | C7H2F7N | 233.089 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24326.06
also known as B24326-06
Price (USD)
180.65
Online Exclusive
201.00
Save 20.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
180.65
Online Exclusive
201.00
Save 20.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS651-83-2
IUPAC Name2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
Molecular FormulaC7H2F7N
InChI KeyFJOACTZFMHZHSC-UHFFFAOYSA-N
SMILESNC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4290-1.4330 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only