3-Methoxyphenyl isocyanate, 99%
3-Methoxyphenyl isocyanate, 99%
3-Methoxyphenyl isocyanate, 99%
3-Methoxyphenyl isocyanate, 99%
Thermo Scientific Chemicals

3-Methoxyphenyl isocyanate, 99%

CAS: 18908-07-1 | C8H7NO2 | 149.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24329.14
also known as B24329-14
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS18908-07-1
IUPAC Name1-isocyanato-3-methoxybenzene
Molecular FormulaC8H7NO2
InChI KeyNPOVTGVGOBJZPY-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.5420-1.5450 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only