2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab.
2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab.
2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab.
Thermo Scientific Chemicals

2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab.

CAS: 45115-53-5 | C7H7F5O2 | 218.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24340.06
also known as B24340-06
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.70
Save 12.05 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.70
Save 12.05 (14%)
Each
Chemical Identifiers
CAS45115-53-5
IUPAC Name2,2,3,3,3-pentafluoropropyl 2-methylprop-2-enoate
Molecular FormulaC7H7F5O2
InChI KeyCLISWDZSTWQFNX-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OCC(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3455-1.4505 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only