2-(Trifluoromethyl)benzeneboronic acid, 97%
2-(Trifluoromethyl)benzeneboronic acid, 97%
2-(Trifluoromethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzeneboronic acid, 97%

CAS: 1423-27-4 | C7H6BF3O2 | 189.928 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24343.14
also known as B24343-14
Price (USD)
435.65
Online Exclusive
484.00
Save 48.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
435.65
Online Exclusive
484.00
Save 48.35 (10%)
Each
Chemical Identifiers
CAS1423-27-4
IUPAC Name[2-(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC7H6BF3O2
InChI KeyJNSBEPKGFVENFS-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
Assay (Aqueous acid-base Titration)≥96.0%
CommentContains varying amounts of anhydride
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only