4-Bromobenzamide, 97%
4-Bromobenzamide, 97%
4-Bromobenzamide, 97%
Thermo Scientific Chemicals

4-Bromobenzamide, 97%

CAS: 698-67-9 | C7H6BrNO | 200.035 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number B24348.18
also known as B24348-18
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
293.00
Save 43.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
293.00
Save 43.35 (15%)
Each
Chemical Identifiers
CAS698-67-9
IUPAC Name4-bromobenzamide
Molecular FormulaC7H6BrNO
InChI KeyZRWNRAJCPNLYAK-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)187.0-196.0?C
Assay (HPLC)≥96.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only