Dimethyl glutarate, 98%
Dimethyl glutarate, 98%
Dimethyl glutarate, 98%
Thermo Scientific Chemicals

Dimethyl glutarate, 98%

CAS: 1119-40-0 | C7H12O4 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B24359.14
also known as B24359-14
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.80
Save 5.15
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.80
Save 5.15
Each
Chemical Identifiers
CAS1119-40-0
IUPAC Name1,5-dimethyl pentanedioate
Molecular FormulaC7H12O4
InChI KeyXTDYIOOONNVFMA-UHFFFAOYSA-N
SMILESCOC(=O)CCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4220-1.4260 @ 20?C
FormLiquid
Dimethyl glutarate is applied as paint and graffiti removers, nail polish removers, hand cleaners, adhesives, sealants and caulks, automotive cleaning products, bathroom and tile cleaners, general purpose cleaners, spot removers, floor maintenance products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl glutarate is applied as paint and graffiti removers, nail polish removers, hand cleaners, adhesives, sealants and caulks, automotive cleaning products, bathroom and tile cleaners, general purpose cleaners, spot removers, floor maintenance products.

Solubility
Soluble in water. (53 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhong Yin Zhang.; Jack E. Dixon. Active site labeling of the Yersinia protein tyrosine phosphatase: The determination of the pKa of the active site cysteine and the function of the conserved histidine 402.Biochemistry. 1993, 32 (36),9340-9345 .
  2. Joseph Wang.; Mustafa Musameh. Carbon nanotube screen-printed electrochemical sensors. Analyst. 2004, 129 (1), 1-2 .