2,4-Dimethoxybenzeneboronic acid, 98%
2,4-Dimethoxybenzeneboronic acid, 98%
2,4-Dimethoxybenzeneboronic acid, 98%
Thermo Scientific Chemicals

2,4-Dimethoxybenzeneboronic acid, 98%

CAS: 133730-34-4 | C8H11BO4 | 181.982 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24374.03
also known as B24374-03
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.50
Save 7.85 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.50
Save 7.85 (15%)
Each
Chemical Identifiers
CAS133730-34-4
IUPAC Name(2,4-dimethoxyphenyl)boronic acid
Molecular FormulaC8H11BO4
InChI KeySQTUYFKNCCBFRR-UHFFFAOYSA-N
SMILESCOC1=CC=C(B(O)O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5%
Assay (Aqueous acid-base Titration)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only