(+/-)-2-Methylpiperazine, 98%
(+/-)-2-Methylpiperazine, 98%
(+/-)-2-Methylpiperazine, 98%
Thermo Scientific Chemicals

(+/-)-2-Methylpiperazine, 98%

CAS: 109-07-9 | C5H12N2 | 100.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B24405.14
also known as B24405-14
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Chemical Identifiers
CAS109-07-9
IUPAC Name2-methylpiperazine
Molecular FormulaC5H12N2
InChI KeyJOMNTHCQHJPVAZ-UHFFFAOYNA-N
SMILESCC1CNCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1.0%
Appearance (Color)White to yellow
FormCrystalline powder or crystals and/or chunks
Melting Point (clear melt)61-67°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only