4-tert-Butylbenzeneboronic acid, 97%
4-tert-Butylbenzeneboronic acid, 97%
4-tert-Butylbenzeneboronic acid, 97%
Thermo Scientific Chemicals

4-tert-Butylbenzeneboronic acid, 97%

CAS: 123324-71-0 | C10H15BO2 | 178.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24408.03
also known as B24408-03
Price (USD)
26.65
Online exclusive
29.40
Save 2.75
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
26.65
Online exclusive
29.40
Save 2.75
Each
Chemical Identifiers
CAS123324-71-0
IUPAC Name(4-tert-butylphenyl)boronic acid
Molecular FormulaC10H15BO2
InChI KeyMNJYZNVROSZZQC-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥96.0%
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only