3-Methoxybenzeneboronic acid, 97%
3-Methoxybenzeneboronic acid, 97%
3-Methoxybenzeneboronic acid, 97%
3-Methoxybenzeneboronic acid, 97%
Thermo Scientific Chemicals

3-Methoxybenzeneboronic acid, 97%

CAS: 10365-98-7 | C7H9BO3 | 151.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24412.03
also known as B24412-03
Price (USD)
32.65
Online Exclusive
35.60
Save 2.95 (8%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
35.60
Save 2.95 (8%)
Each
Add to cart
Chemical Identifiers
CAS10365-98-7
IUPAC Name(3-methoxyphenyl)boronic acid
Molecular FormulaC7H9BO3
InChI KeyNLLGFYPSWCMUIV-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale gray
Assay (Aqueous acid-base Titration)≥96.0%
Melting Point (clear melt)154.0-164.0?C
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only