1-Tridecene, 97%
1-Tridecene, 97%
1-Tridecene, 97%
Thermo Scientific Chemicals

1-Tridecene, 97%

CAS: 2437-56-1 | C13H26 | 182.351 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24413.06
also known as B24413-06
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.90
Save 7.25 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.90
Save 7.25 (14%)
Each
Chemical Identifiers
CAS2437-56-1
IUPAC Nametridec-1-ene
Molecular FormulaC13H26
InChI KeyVQOXUMQBYILCKR-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4325-1.4395 @ 20°C
It is used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Sara M. Aschmann. ; Roger Atkinson. Rate constants for the gas-phase reactions of OH radicals with E-7-tetradecene, 2-methyl-1-tridecene and the C7-C14 1-alkenes at 295 ± 1 K. Phys. Chem. Chem. Phys. 2008, 10 (28),4159-4164 .
  2. Kazuhiko Takai..; Tadahiro Kakiuchi.; Yasutaka Kataok.; Kiitiro Utimoto. A Novel Catalytic Effect of Lead on the Reduction of a Zinc Carbenoid with Zinc Metal Leading to a Geminal Dizinc Compound. Acceleration of the Wittig-Type Olefination with the RCHX2-TiCl4-Zn Systems by Addition of Lead. J. Org. Chem. 1994, 59 (10),2668-2670 .