1,2-Diiodobenzene, 98%
1,2-Diiodobenzene, 98%
1,2-Diiodobenzene, 98%
Thermo Scientific Chemicals

1,2-Diiodobenzene, 98%

CAS: 615-42-9 | C6H4I2 | 329.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24425.14
also known as B24425-14
Price (USD)
382.65
Special offer
Online exclusive
Ends: 30-Jun-2026
450.00
Save 67.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
382.65
Special offer
Online exclusive
Ends: 30-Jun-2026
450.00
Save 67.35 (15%)
Each
Chemical Identifiers
CAS615-42-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to yellow
Assay (GC)>97.5%
Formliquid
Refractive Index> 1.7100
1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.

Solubility
Slightly miscible with water.

Notes
Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatibile with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Akehi, M.; Kawamoto, M.; Mandai, T. Chiral 1-(1,3-dithian-2-yl) prop-2-en-1-ols: new scaffolds for enantiopure alpha-hydroxyaldehydes. Tetrahedron 2015, 71 (37), 6488-6498.
  2. Nagarjuna, G.; Ren, Y.; Moore, J. S. Synthesis and reactivity of anthracenyl-substituted arenediynes. Tetrahedron Lett. 2015, 56 (23), 3155-3159.