beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%
beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%
beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%
Thermo Scientific Chemicals

beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%

CAS: 13035-61-5 | C13H18O9 | 318.278 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24426.06
also known as B24426-06
Price (USD)
39.65
Online Exclusive
44.00
Save 4.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
44.00
Save 4.35 (10%)
Each
Chemical Identifiers
CAS13035-61-5
IUPAC Name[3,4,5-tris(acetyloxy)oxolan-2-yl]methyl acetate
Molecular FormulaC13H18O9
InChI KeyIHNHAHWGVLXCCI-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)79.0-85.0?C
FormCrystals or powder or crystalline powder
Optical Rotation-12.7 ± 0.3? (C=2.4 in chloroform)
Assay (GC)≥98.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only