3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol
3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol
3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol
Thermo Scientific Chemicals

3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol

CAS: 2039-86-3 | C8H7Br | 183.048 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24497.03
also known as B24497-03
Price (USD)
41.65
Online Exclusive
46.50
Save 4.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
46.50
Save 4.85 (10%)
Each
Chemical Identifiers
CAS2039-86-3
IUPAC Name1-bromo-3-ethenylbenzene
Molecular FormulaC8H7Br
InChI KeyKQJQPCJDKBKSLV-UHFFFAOYSA-N
SMILESBrC1=CC=CC(C=C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5875-1.5925 @ 20?C
Solution Test10% in methanol: clear
Appearance (Color)Clear colorless to pale yellow
Solution Test10% in diethyl ether: clear
Stabilizer0.1% 4-tert-butylcatechol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only