Methyl benzenesulfonate, 98%
Methyl benzenesulfonate, 98%
Methyl benzenesulfonate, 98%
Methyl benzenesulfonate, 98%
Thermo Scientific Chemicals

Methyl benzenesulfonate, 98%

CAS: 80-18-2 | C7H8O3S | 172.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B24515.14
also known as B24515-14
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.20
Save 7.55 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.20
Save 7.55 (15%)
Each
Add to cart
Chemical Identifiers
CAS80-18-2
IUPAC Namemethyl benzenesulfonate
Molecular FormulaC7H8O3S
InChI KeyCZXGXYBOQYQXQD-UHFFFAOYSA-N
SMILESCOS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5150-1.5190 @ 20?C
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.

Solubility
Miscible with chloroform, methanol, ethanol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

General References:

  1. Alkylating agent.
  2. Giricheva, N. I.; Fedorov, M. S.; Girichev, G. V. Conformations of methylbenzenesulfonate and its substituted derivatives: gas-phase electron diffraction versus vibrational spectroscopy. Struct. Chem. 2015, 26 (5), 1543-1553.
  3. Zhang, X.; Jiang, X.; Li, Y.; Lin, Z.; Zhang, G.; Wu, Y. Synthesis, crystal structures and properties of new quinolinium derivatives. Chem. Phys. Lett. 2015, 641, 141-145.