4-Hydroxybenzoylacrylic acid, 97%
4-Hydroxybenzoylacrylic acid, 97%
4-Hydroxybenzoylacrylic acid, 97%
Thermo Scientific Chemicals

4-Hydroxybenzoylacrylic acid, 97%

CAS: 24849-48-7 | C10H7O4 | 191.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24519.14
also known as B24519-14
Price (USD)
753.65
Online Exclusive
837.00
Save 83.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
753.65
Online Exclusive
837.00
Save 83.35 (10%)
Each
Chemical Identifiers
CAS24849-48-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only