2,6-Dimethylpiperidine, predominantly cis, 99%
2,6-Dimethylpiperidine, predominantly cis, 99%
2,6-Dimethylpiperidine, predominantly cis, 99%
Thermo Scientific Chemicals

2,6-Dimethylpiperidine, predominantly cis, 99%

CAS: 504-03-0 | C7H15N | 113.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number B24524.18
also known as B24524-18
Price (USD)
68.70
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
68.70
Each
Chemical Identifiers
CAS504-03-0
IUPAC Name(2R,6S)-2,6-dimethylpiperidin-1-ium
Molecular FormulaC7H16N
InChI KeySDGKUVSVPIIUCF-KNVOCYPGSA-O
SMILESC[C@H]1CCC[C@@H](C)[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4385-1.4415 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only