Eosin Yellowish
Eosin Yellowish
Eosin Yellowish
Thermo Scientific Chemicals

Eosin Yellowish

CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B24535.18
also known as B24535-18
Price (USD)
46.90
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
46.90
Each
Chemical Identifiers
CAS17372-87-1
IUPAC Namedisodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6Br4Na2O5
InChI KeySEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Loss on Drying≤10.0%
Appearance (Color)Red to brown
Assay (Dye)≥90.0% (on dry substance)
Eosin Yellowish noted to reduce the affinity of Mg2+-ATPase for ATP and increase the enzyme affinity for Mg2+ at concentrations of 10-50 M. At higher concentrations near 100 ?M the affinity for Mg2+-ATPase for the ion-activator has been shown to be reduced. This compound has also been reported to be a photosensitizer that works as a molecular photoelectrode via catalyzation of the visible-light-driven electron-transfer reaction. Eosin Y is also used to stain structures such as muscle, cytoplasm, and collagen. This agent also enhances fluorescent quenching abilities by forming a complex with polyvinulpyrrolidone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eosin Yellowish noted to reduce the affinity of Mg2+-ATPase for ATP and increase the enzyme affinity for Mg2+ at concentrations of 10-50 μM. At higher concentrations near 100 μM the affinity for Mg2+-ATPase for the ion-activator has been shown to be reduced. This compound has also been reported to be a photosensitizer that works as a molecular photoelectrode via catalyzation of the visible-light-driven electron-transfer reaction. Eosin Y is also used to stain structures such as muscle, cytoplasm, and collagen. This agent also enhances fluorescent quenching abilities by forming a complex with polyvinulpyrrolidone.

Solubility
Soluble in water, fairly soluble in alcohol, insoluble in ether

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A.K. Golder.; N. Hridaya.; A.N. Samanta.; S. Ray. Electrocoagulation of methylene blue and eosin yellowish using mild steel electrodes. Journal of Hazardous Materials. 2005, 127(1-3), 134-140.
  2. Yuko Ohashi.; Makoto Matsuoka. Synthesis of Stress Proteins in Tobacco Leaves. Plant Cell Physiol. 1985, 26(3), 473-480..
  3. Adsorption and fluorescence indicator.