Methyl phenylpropiolate, 97%
Methyl phenylpropiolate, 97%
Methyl phenylpropiolate, 97%
Thermo Scientific Chemicals

Methyl phenylpropiolate, 97%

CAS: 4891-38-7 | C10H8O2 | 160.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24587.06
also known as B24587-06
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.30
Save 9.65 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.30
Save 9.65 (14%)
Each
Chemical Identifiers
CAS4891-38-7
IUPAC Namemethyl 3-phenylprop-2-ynoate
Molecular FormulaC10H8O2
InChI KeyJFGWPXKGINUNDH-UHFFFAOYSA-N
SMILESCOC(=O)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5595-1.5645 @ 20?C
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only