5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%
Thermo Scientific Chemicals

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%

CAS: 1046-56-6 | C20H14N4 | 310.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24601.14
also known as B24601-14
Price (USD)
361.65
Special offer
Online exclusive
Ends: 30-Jun-2026
425.00
Save 63.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
361.65
Special offer
Online exclusive
Ends: 30-Jun-2026
425.00
Save 63.35 (15%)
Each
Chemical Identifiers
CAS1046-56-6
IUPAC Name5,6-diphenyl-3-(pyridin-2-yl)-1,2,4-triazine
Molecular FormulaC20H14N4
InChI KeyOTMYLOBWDNFTLO-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)189.0-195.0?C
Appearance (Color)Yellow
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
View more
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Reagent for the determination of Cu, and for the spectrophotometric determination of Fe(II) and Ru(II): Anal. Chem., 46, 992 (1974); Anal. Chim. Acta, 78, 423 (1975).
  2. Yamada, M.; Muto, Y.; Kurihara, H.; Slanina, Z.; Suzuki, M.; Maeda, Y.; Rubin, Y.; Olmstead, M. M.; Balch, A. L.; Nagase, S.; Lu, X.; Akasaka, T. Regioselective Cage Opening of La2@D2(10611)-C72 with 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine. Angew. Chem. Int. Ed. 2015, 54 (7), 2232-2235.
  3. Anjomshoa, M.; Hadadzadeh, H.; Mahani, M. T.; Fatemi, S. J.; Sardou, M. A.; Rudbari, H. A.; Nardo, V. M. A mononuclear Cu(II) complex with 5,6-diphenyl-3-(2-pyridyl)-1,2,4-triazine: Synthesis, crystal structure, DNA- and BSA-binding, molecular modeling, and anticancer activity against MCF-7, A-549, and HT-29 cell lines. Angew. Chem. Int. Ed. 2015, 96, 66-82.